Order Entry
China
ContactUsLinkComponent
Cryogenic Vials,2.0ml,Self-standing,Brown and non-transparent,PP,Working Temperature from -86°C to 121°C Sterile, DNase&Rnase free 1 * 2.000 items
Catalog # BGIX81-7205
Supplier:  BIOLOGIX
Cryogenic Vials,2.0ml,Self-standing,Brown and non-transparent,PP,Working Temperature from -86°C to 121°C Sterile, DNase&Rnase free 1 * 2.000 items
Catalog # BGIX81-7205
Supplier:  BIOLOGIX