VWR® Basic Mob Caps, Double Elastic
Orange, 530 mm
Catalog # VWRI113-8867
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable pleated caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Double elastic
- Available in a wide range of colours
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.
Documents
CoC_VWRI113-8863_D9190147 CoC_VWRI113-8863_D9201717 CoC_VWRI113-8863_D9210531 COC_VWRI113-8863_D9220426 COC_VWRI113-8863_D9221000
More
Food Contact Certificate
Additional Product Details
TDS Mob Caps, Double Elastic
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Ø:530 mm
- Colour:Orange
- Seq:06



