Order Entry
China
Orders LinkContactUsLinkComponent
VWR®, Sampling Scoops

VWR®, Sampling Scoops

50 ml, 161 mm, White
Catalog # VWRU10820-349
VWR®, Sampling Scoops
Catalog # VWRU10820-349
VWR®, Sampling Scoops
VWR®, Sampling Scoops

50 ml, 161 mm, White

Price
¥5,718.00
Log in for availability
1 * 100 件
Product Details & Documents

PS, white. These sterile scoops are ideal for sampling powder, granules and pastes.

  • Individually packed
  • Sterilised by gamma radiation

Thanks to the special handle construction, and the slightly angled edge, the scoop stands horizontally and can be laid down without the material trickling out.

Certifications: Production, assembly and packaging according to cleanroom Class 7 (10000). According to EU food and FDA guidelines.

Caution: *The product(s) below are outside the scope of the EU Directive 2019/904.

Specifications
  • Capacity:
    50 ml
  • Colour:
    White
  • Length:
    161 mm
  • Seq:
    03