¥0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
China
ContactUsLinkComponent
Matrix 0.75ml 2D,聚丙烯,V底管,大包装 1 * 1.000 件
MATR3730
:
THERMO MATRIX TECHNOLOGIES
Matrix 0.75ml 2D,聚丙烯,V底管,大包装 1 * 1.000 件
MATR3730
:
THERMO MATRIX TECHNOLOGIES