¥0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
China
ContactUsLinkComponent
Matrix V底管 Trakmates 2D Bar 0.75mL管未灭菌 1 * 960 件
Catalog # MATR3731
Supplier:
THERMO MATRIX TECHNOLOGIES
Expand image
Matrix V底管 Trakmates 2D Bar 0.75mL管未灭菌 1 * 960 件
Catalog # MATR3731
Supplier:
THERMO MATRIX TECHNOLOGIES