Order Entry
China
Orders LinkContactUsLinkComponent
Measuring scoops

Measuring scoops

50 ml, 199 mm, 35 mm
Catalog # SAMP1900A-50
Supplier:   SAMPLING SYSTEMS
Measuring scoops
Catalog # SAMP1900A-50
Supplier:  SAMPLING SYSTEMS
Measuring scoops
Measuring scoops

50 ml, 199 mm, 35 mm

Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These measuring scoops made from high quality 316 stainless steel are ideal for scooping out fixed volumes of powder.

  • Crevice-free
  • Polished
Specifications
  • Ø:
    35 mm
  • Capacity:
    50 ml
  • Length:
    199 mm
  • Seq:
    05

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.