Order Entry
China
ContactUsLinkComponent
Compact, self-priming polyethylene lever pump, suitable for alkaline liquids and many acids. The telescope suction tube for lever pump is steplessly extendable to 914 mm. Pump capacity is about 300 mL per stroke. With integrated barrel screw connecti 1 * 1 items
Catalog # BURK5613-0020
Supplier:  BURKLE
Compact, self-priming polyethylene lever pump, suitable for alkaline liquids and many acids. The telescope suction tube for lever pump is steplessly extendable to 914 mm. Pump capacity is about 300 mL per stroke. With integrated barrel screw connecti 1 * 1 items
Catalog # BURK5613-0020
Supplier:  BURKLE