Order Entry
China
ContactUsLinkComponent
Anti-Rabbit Detection Module for Wes, Peggy Sue or Sally Sue Use with a Wes or Peggy Sue/Sally Sue Seperation Module. Includes: Luminol-S, Preoxide, Antibody Diluent 2, Anti-Rabbit Secondary Antibody and Streptavidin-HR
Catalog # PRTSDM-001
Supplier:  PROTEIN SIMPLE
Anti-Rabbit Detection Module for Wes, Peggy Sue or Sally Sue Use with a Wes or Peggy Sue/Sally Sue Seperation Module. Includes: Luminol-S, Preoxide, Antibody Diluent 2, Anti-Rabbit Secondary Antibody and Streptavidin-HR
Catalog # PRTSDM-001
Supplier:  PROTEIN SIMPLE