Plastic feeding tubes, 15ga x 100mm. 50 sterile pouches of 5. Polypropylene female luer (blue) and flexible polypropylene tube with soft elastomer tip. For rodent oral gavage. Disposable. Designed to minimize trauma to esophagus. 1 * 250 ST
Catalog # MSPPFTP-15-100-250
Supplier: MARKET SOURCE PART PROCESS