Order Entry
China
ContactUsLinkComponent
Bottletop dispenser Dispensette® S, Analog-adjustable, DE-M marking, 10-100 ml, w. recirculation valve 1 * 1 item
Catalog # BRND4600171-AT
Supplier:  BRAND
Bottletop dispenser Dispensette® S, Analog-adjustable, DE-M marking, 10-100 ml, w. recirculation valve 1 * 1 item
Catalog # BRND4600171-AT
Supplier:  BRAND