Order Entry
China
ContactUsLinkComponent
Bottletop dispenser Dispensette® S Organic, Analog-adjustable, DE-M marking, 1-10 ml, with recirculation valve 1 * 1 item
Catalog # BRND4630141-AT
Supplier:  BRAND
Bottletop dispenser Dispensette® S Organic, Analog-adjustable, DE-M marking, 1-10 ml, with recirculation valve 1 * 1 item
Catalog # BRND4630141-AT
Supplier:  BRAND