Order Entry
China
ContactUsLinkComponent
Stainless steel bottom drip tray, 15 mm rim (may affect the temperature distribution) – cannot be used in connection with option K1 MAX. LOADING 3 KG 1 * 1 items
Catalog # MEMMB04359UF160TS
Supplier:  MEMMERT
Stainless steel bottom drip tray, 15 mm rim (may affect the temperature distribution) – cannot be used in connection with option K1 MAX. LOADING 3 KG 1 * 1 items
Catalog # MEMMB04359UF160TS
Supplier:  MEMMERT