Order Entry
China
ContactUsLinkComponent
ENTRY PORT 23 MM CLEAR DIAMETER FOR INTRODUCING CONNECTIONS AT THE SIDE CAN BE CLOSED BY FLAP STANDARD POSITIONS 1 * 1 items
Catalog # MEMMUF160TSF3
Supplier:  MEMMERT
ENTRY PORT 23 MM CLEAR DIAMETER FOR INTRODUCING CONNECTIONS AT THE SIDE CAN BE CLOSED BY FLAP STANDARD POSITIONS 1 * 1 items
Catalog # MEMMUF160TSF3
Supplier:  MEMMERT