Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PH7缓冲液20ML, PK20 1 * 400 毫升
Catalog # BDHBBDH50440-20PK
Supplier:
BDH US PRODUCED
Expand image
PH7缓冲液20ML, PK20 1 * 400 毫升
Catalog # BDHBBDH50440-20PK
Supplier:
BDH US PRODUCED