Order Entry
China
ContactUsLinkComponent
Automatic burette, Pellet pattern, w/o intermediate stopcock, w/o bottle, BLAUBRAND® class AS, DE-M marking, 25 ml:0,05 ml Boro 3.3, Schellbach lateral needle-valve stopcock PTFE-key, grad. in blue, individual certificate 1 * 1 items
  BRND922522
 :  BRAND
Automatic burette, Pellet pattern, w/o intermediate stopcock, w/o bottle, BLAUBRAND® class AS, DE-M marking, 25 ml:0,05 ml Boro 3.3, Schellbach lateral needle-valve stopcock PTFE-key, grad. in blue, individual certificate 1 * 1 items
  BRND922522
 :  BRAND