Order Entry
China
Orders LinkContactUsLinkComponent
Carts
Carts
Starsys cart with interchangeable drawers, color-coded drawer pulls and Label holders available to quickly identify drawer contents, 578×632×1060 mm
Catalog # METOSXRSBED
Supplier:   METRO
Carts
Catalog # METOSXRSBED
Supplier:  METRO
Carts
Carts
Starsys cart with interchangeable drawers, color-coded drawer pulls and Label holders available to quickly identify drawer contents, 578×632×1060 mm
Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents
The Starsys carts are available in a wide variety of heights and system widths and are ideal for the storage needs in the laboratories. The carts features smooth polymer top/worksurface and are provided with 127 mm polymer swivel casters, the front two casters comes with total-lock toe brake to lock both the wheel and the horn.
  • Easy to clean
  • Interchangeable drawers (76, 152 and 230 mm) with removable totes can be fully extended for easy access
  • Colour-coded drawer pulls are available in an array of choices
  • Label holders available to quickly identify drawer contents
规格
  • Description:
    Starsys cart with interchangeable drawers, color-coded drawer pulls and Label holders available to quickly identify drawer contents
  • W×D×H:
    578×632×1060 mm