Biotix variable volume 8-ch Pipettor 20 to 300µl, Soft spring system for extremely low pipetting forces, Controlled volume setting t o prevent accidental volume changes, Fully autoclavable (120°C/0.1MPa/20 min), A unique serial number, Finger hook: t 1 * 1 items
BITXBTX-8X-300
: BIOTIX