Order Entry
China
Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Conductivity standard, 84 µS/cm, 10
Catalog # METT30111140
Supplier:   METTLER TOLEDO
Conductivity standards
Catalog # METT30111140
Supplier:  METTLER TOLEDO
CAS Number:  
Conductivity standards
Conductivity standards
Conductivity standard, 84 µS/cm, 10
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 件
Product Details & Documents
A slight change in temperature usually has a big impact on the conductivity value of a standard solution. A table on every bottle label indicates the conductivity values at the most common measurement temperatures.
During calibration the meter automatically references to this table for temperature compensation. If possible, calibration and measurements should be performed at the same temperature
Documents
Certificates
Search by Lot Number
Specifications
Specifications
  • Pk:
    10
  • Description:
    Conductivity standard
  • Conductivity:
    84 µS/cm
Specifications