VWR®, Dissecting forceps
Curved, serrated, fine tips, 152 mm
VWRU82027-406_U
Dissecting forceps made of stainless steel. Available straight or curved and in different lengths.
- Type:Curved, serrated, fine tips
- Length:152 mm
- Seq:12
Dissecting forceps made of stainless steel. Available straight or curved and in different lengths.