Order Entry
China
ContactUsLinkComponent
Liquidproof gloves made of natural latex blended with neoprene and nitrile. Length 36 cm, thickness 0,5 mm colour: orange, Unit = 12 pairs Anatomical form Chlorinated internal surface. Finished, tested and packaged in certified Class 100 (M3.5) clean 1 * 12 PAIR
Catalog # MAPA518567
Supplier:  MAPA
Liquidproof gloves made of natural latex blended with neoprene and nitrile. Length 36 cm, thickness 0,5 mm colour: orange, Unit = 12 pairs Anatomical form Chlorinated internal surface. Finished, tested and packaged in certified Class 100 (M3.5) clean 1 * 12 PAIR
Catalog # MAPA518567
Supplier:  MAPA