Order Entry
China
ContactUsLinkComponent
Graduated pipette, BLAUBRAND® class A conf.cert., 0,2 ml:0,002 ml cal. to contain AR-Glas, with individual certificate 1 * 1 items
Catalog # BRND927704
Supplier:  BRAND
undefined
Graduated pipette, BLAUBRAND® class A conf.cert., 0,2 ml:0,002 ml cal. to contain AR-Glas, with individual certificate 1 * 1 items
Catalog # BRND927704
Supplier:  BRAND