Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
pH Buffer,pH4.01,red,20mL*20 1 * 20 items
Catalog # BDHB5016-20ML
Supplier:
BDH US PRODUCED
Expand image
pH Buffer,pH4.01,red,20mL*20 1 * 20 items
Catalog # BDHB5016-20ML
Supplier:
BDH US PRODUCED