Order Entry
China
ContactUsLinkComponent
DURAN Group Dispenser adjustable volume 1.0-10.0 ml, Scope of delivery: three PP threaded adapters, telescopic intake tube (200–350 mm), assembly key, instructions for use, 1 * 1 items
Catalog # SCOT291150908
Supplier:  DWK LIFE SCIENCES
undefined
DURAN Group Dispenser adjustable volume 1.0-10.0 ml, Scope of delivery: three PP threaded adapters, telescopic intake tube (200–350 mm), assembly key, instructions for use, 1 * 1 items
Catalog # SCOT291150908
Supplier:  DWK LIFE SCIENCES