Order Entry
China
Orders LinkContactUsLinkComponent
Lockout hasps

Lockout hasps

Lockout hasp, 25 mm diameter jaws, Red
Catalog # BRDY65375
Supplier:   BRADY
Lockout hasps
Catalog # BRDY65375
Supplier:  BRADY
Lockout hasps
Lockout hasps

Lockout hasp, 25 mm diameter jaws, Red

Price
Inquire For Price
Log in for availability
1 * 12 件
Product Details & Documents

These lockout hasps are ideal for use during group lockout situations.

  • Hasps are constructed of vinyl-coated high tensile steel and are plated to resist rust
  • Lock holes are 9,5 mm (³/₈") diameter to accommodate most standard lock shackles
Specifications
  • Type:
    Lockout hasp, 25 mm diameter jaws
  • Colour:
    Red
  • Seq:
    01