Formic acid
Glass bottle, 250 mL
Catalog # CHLCR01996320023
Supplier: LOCAL DEALS
Jump to:
Product Details & Documents
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Boiling Pt:100…101 °C (1013 hPa)
- Melting Pt:8.2…8.4 °C
- Density:1.22 g/cm³ (25 °C)
- Flash Pt:48 °C
- EINECS:200-579-1
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- Storage Temperature:Ambient
- UN:1779
- REACH:01-2119491174-37
Specifications
Specifications
- Pack type:Glass bottle
- Pk:250 mL



