Plastic feeding tubes, 20ga x 30mm. 50 sterile pouches of 5. Polypropylene female luer (yellow) and flexible polypropylene tube with soft elastomer tip. For rodent oral gavage. Disposable. Designed to minimize trauma to esophagus. 1 * 250 items
Catalog # MSPPFTP-20-30
Supplier: MARKET SOURCE PART PROCESS