Order Entry
China
ContactUsLinkComponent
MINUS 47 TO 100, 12L, INCLUDES DRAIN, RELAY DIGITAL BATH WITH POWERFUL MULTI-STAGE PUMP, USB, AND PROGRAMMABILITY 1 * 1 items
Catalog # GRANTXF200-R5
Supplier:  GRANT INSTRUMENTS
MINUS 47 TO 100, 12L, INCLUDES DRAIN, RELAY DIGITAL BATH WITH POWERFUL MULTI-STAGE PUMP, USB, AND PROGRAMMABILITY 1 * 1 items
Catalog # GRANTXF200-R5
Supplier:  GRANT INSTRUMENTS