Affinity chromatography columns, HiTrap™ Protein L
Product Details & Documents
Columns prepacked with Capto™ L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.
- Prepacked for easy screening and process development, as well as small scale purification
- Columns easily connected in series for increased capacity
- Designed for use with syringe, peristalic pump, or chromatography system
Documents
Specifications
- Particle Size:85 µm
- Capacity:5×1 ml
- Description:HiTrap™ Protein L
- Seq:03
- Column hardware:PP
- Bed dimensions:7×25 mm
- Flow velocity:<500 cm/h
- Cat. No.:GEHE17-5478-51
- Max. pressure:5 bar
- Storage:2 to 8 °C, 20% Ethanol
- pH range:2 - 10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



