0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Balance Precision NewClassic MS16001L 16200g x 0.1g with Internal Cal 1 * 1 SET
METBMS16001L
:
METTLER TOLEDO
Balance Precision NewClassic MS16001L 16200g x 0.1g with Internal Cal 1 * 1 SET
METBMS16001L
:
METTLER TOLEDO