Order Entry
China
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

pH buffer standard solution with certificate of analysis, 125 ml, 7,00, 1
Catalog # HACHLZW9461.97
Supplier:   HACH
pH buffer solutions
Catalog # HACHLZW9461.97
Supplier:  HACH
CAS Number:  
pH buffer solutions
pH buffer solutions

pH buffer standard solution with certificate of analysis, 125 ml, 7,00, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 件
Product Details & Documents
Specifications

Specifications

  • Pk:
    1
  • Description:
    pH buffer standard solution with certificate of analysis, 125 ml
  • pH value:
    7,00

Specifications