pH buffer solutions, colour coded
Buffer solution, 10,01, ±0,02, Carbonate, Blue, 25 °C, 4 L
Catalog # HACH2283656
Jump to:
Discontinued
Product Details & Documents
For calibration of pH meters
- Colour coded for easy identification
- Accuracy (pH): ±0,02 at 25 °C
: Traceable to NIST.
Specifications
Specifications
- pH buffer type:Carbonate
- Pk:4 L
- Reference temperature:25 °C
- Colour:Blue
- Description:Buffer solution
- pH accuracy:±0,02
- pH value:10,01



