Order Entry
China
Orders LinkContactUsLinkComponent
 

Pipette tips, GripTips

Standard, 5000 µl, +, 145,24 mm, Bulk, 100 tips/bag (individually wrapped)
  INTL4456
Pipette tips, GripTips
  INTL4456
Pipette tips, GripTips
Pipette tips, GripTips

Standard, 5000 µl, +, 145,24 mm, Bulk, 100 tips/bag (individually wrapped)

 
 
   
1 * 100 件
 

GripTips are manufactured from virgin medical grade materials and designed for use with the VIAFLO pipettes. Their features perfectly match the tip fitting to form a complete system. Without hammering, the GripTips literally snap onto the pipette's tip fitting, which prevents tips from falling off.

  • Reduced tip attachment and ejection forces for comfortable, stress-free pipetting
  • Extended length design enables easy access into tubes and deep well blocks
  • Sterile tips are certified free of RNase, DNase, pyrogens, PCR inhibitors and endotoxins
  • GREEN CHOICE refill system: environmentally friendly refill system to reuse the current tip racks and thus reduce plastic waste by 60%

 : Available in bulk packs, tip racks with single inserts in 96 and 384 fold configuration or as GREEN CHOICE refills.

 
  • Packed:
    Bulk, 100 tips/bag (individually wrapped)
  • Sterile:
    +
  • Volume:
    5000 µl
  • Length:
    145,24 mm
  • Seq:
    27
  • Tip style:
    Standard