Order Entry
China
Orders LinkContactUsLinkComponent
 

Pipette tips, GripTips

Standard, 12,5 µl, -, 35,56 mm, Bulk, 1000 tips/bag
Catalog # INTL4411
Supplier:   INTEGRA Biosciences
Pipette tips, GripTips
Catalog # INTL4411
Pipette tips, GripTips
Pipette tips, GripTips

Standard, 12,5 µl, -, 35,56 mm, Bulk, 1000 tips/bag

Price
Inquire For Price
Log in for availability
1 * 1.000 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

GripTips are manufactured from virgin medical grade materials and designed for use with the VIAFLO pipettes. Their features perfectly match the tip fitting to form a complete system. Without hammering, the GripTips literally snap onto the pipette's tip fitting, which prevents tips from falling off.

  • Reduced tip attachment and ejection forces for comfortable, stress-free pipetting
  • Extended length design enables easy access into tubes and deep well blocks
  • Sterile tips are certified free of RNase, DNase, pyrogens, PCR inhibitors and endotoxins
  • GREEN CHOICE refill system: environmentally friendly refill system to reuse the current tip racks and thus reduce plastic waste by 60%

 : Available in bulk packs, tip racks with single inserts in 96 and 384 fold configuration or as GREEN CHOICE refills.

Specifications
  • Packed:
    Bulk, 1000 tips/bag
  • Sterile:
    -
  • Volume:
    12,5 µl
  • Length:
    35,56 mm
  • Seq:
    01
  • Tip style:
    Standard

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.