0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KLARI-FLEX BOTTLE-TOP FI 50 ML 0.22 PES 1 * 24 items
Catalog # WHAT6518-0502
Supplier:
CYTIVA WHATMAN
Expand image
KLARI-FLEX BOTTLE-TOP FI 50 ML 0.22 PES 1 * 24 items
Catalog # WHAT6518-0502
Supplier:
CYTIVA WHATMAN