Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Iron(II) chloride tetrahydrate, ReagentPlus(R), 99% 1 * 250 g
Catalog # HONC220299-250G
Supplier:
HONEYWELL
SDS
Expand image
Iron(II) chloride tetrahydrate, ReagentPlus(R), 99% 1 * 250 g
Catalog # HONC220299-250G
Supplier:
HONEYWELL
SDS
Related Information
Documentation
SDS
sds_220299
Documentation