Sample concentrators, Vivaspin®
Product Details & Documents
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration.
- One-step sample concentration in a single tube for minimal sample handling and reduced sample loss
- Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
- Vertical PES membrane minimises membrane blockage and tolerates high flow rates
- Compatible pH range is from pH 1 to 9
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cutoff (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.
Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3000 to 100000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Documents
Certificates
Search by Lot Number28932218_DATA FILE 28932225_DATA FILE 28932235_DATA FILE 28932236_DATA FILE 28932237_DATA FILE
Datasheet
28932218_HAND BOOK 28932218_INSTRUCTIONS 28932225_HAND BOOK 28932225_INSTRUCTIONS 28932235_HAND BOOK
Manual
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- MWCO:10 kD
- Seq:21
- Filtration area:6,0 cm²
- Cat. No.:GEHE28-9323-60
- Type:Vivaspin® 20
- Dead volume:50 µl
- Ø×H:30×116 mm



