Affinity chromatography columns, HiTrap™ MabSelect™
HiTrap™ MabSelect™ columns are prepacked with MabSelect™, a Bioprocess medium for capuring of monoclonal antibodies(Mabs) from large sample volumes.
- Specificity of binding to the Fc region of IgG is similar to that of native protein A, which allows one-step purification
- Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
- Flow rate:<20 ml/min
- Particle Size:85 µm
- Capacity:1×5 ml
- Description:HiTrap™ MabSelect™
- Seq:02
- Column hardware:PP
- Bed dimensions:16×25 mm
- Cat. No.:GEHE28-4082-55
- Max. pressure:5 bar
- Storage:2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
- pH range:3 - 10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



