跳转到 页眉
跳转到 主要内容
跳转到 页脚
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HEPES SOLUTION BIOXTRA, 1 M, PH 7.0-7.6, STERILE-FILTERED, SUITABLE FOR CELL CULTURE 1 * 20 mL
Catalog # SIALH0887-20ML
Supplier:
SIGMAALDRICH
Certificates
Expand image
HEPES SOLUTION BIOXTRA, 1 M, PH 7.0-7.6, STERILE-FILTERED, SUITABLE FOR CELL CULTURE 1 * 20 mL
Catalog # SIALH0887-20ML
Supplier:
SIGMAALDRICH
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation