Wash buffer, Pierce™
Pierce™ PBS Tween™ 20 buffer, 20X, 7,5, 0,5 L
Catalog # PIER28352
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These wash buffers are designed for use in dialysis, cross-linking, enzyme assays, ELISAs, immunohistochemistry, protein plate-coating, biotinylation applications.
- Ready to use without having to reconstitute with ultrapure water
- Eliminates the possibility of powder remaining in a packet
- 10X or 20X concentration eliminates time spent waiting for powder to dissolve
- Storage as concentrated stock minimizes bench space needed for large volume solutions
Specifications
Specifications
- Pk:0,5 L
- Description:Pierce™ PBS Tween™ 20 buffer, 20X
- pH value:7,5



