Dropping funnel, 10ML, cylindrical, PTFE plug, with pressure equalizing tube, borosilicate glass 3.3. With NS-socket and interchangeable PE stopper, plug with retaining device. With graduations for reading approx. volume. 1 * 1 items
Catalog # LENZ04371311
Supplier: LENZ LABORGLAS