Order Entry
China
Orders LinkContactUsLinkComponent
Buffer solutions, ready to use

Buffer solutions, ready to use

9,2, ±0,02, Borate, Colourless, Plastic bottle, 500 mL
Catalog # ACRO383875000
Buffer solutions, ready to use
Catalog # ACRO383875000
CAS Number:  
Buffer solutions, ready to use

9,2, ±0,02, Borate, Colourless, Plastic bottle, 500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 毫升
Product Details & Documents
  • Traceable to NIST
  • Clear colourless solutions
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    500 mL
  • Colour:
    Colourless
  • pH accuracy:
    ±0,02
  • pH value:
    9,2
  • Cat. No.:
    ACRO383875000
  • Density:
    1

Specifications