¥0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
China
Orders Link
ContactUsLinkComponent
Buffers
pH Calibration Buffers
Buffer solutions, ready to use
Buffer solutions, ready to use
7,00, ±0,02, Phosphate, Colourless, Plastic bottle, 1 L
ACRO383850010
:
Thermo Fisher Scientific
Buffer solutions, ready to use
ACRO383850010
:
Thermo Fisher Scientific
:
Buffer solutions, ready to use
7,00, ±0,02, Phosphate, Colourless, Plastic bottle, 1 L
:
1 * 1 升
-
+
Traceable to NIST
Clear colourless solutions
Documents
SDS
ACRO38383
ACRO38385
ACRO38387
Pack type:
Plastic bottle
pH buffer type:
Phosphate
Pk:
1 L
Colour:
Colourless
pH accuracy:
±0,02
pH value:
7,00
Cat. No.:
ACRO383850010
Density:
1