Order Entry
China
Orders LinkContactUsLinkComponent
Spoon spatulas

Spoon spatulas

210 mm, 29 mm
Catalog # USBE3348
Supplier:   USBECK LABORGERATE
Spoon spatulas
Catalog # USBE3348
Supplier:  USBECK LABORGERATE
Spoon spatulas
Spoon spatulas

210 mm, 29 mm

Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Stainless steel.

  • Double-ended: Spatula and spoon end
  • Spoon width: 29 mm
Specifications
  • Length:
    210 mm
  • Spoon size:
    29 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.