Order Entry
China
ContactUsLinkComponent
GLOVES ALPHATEC. : LIQUID-PROOF NITRILE GLOVE WITH WET GRIP SURFACE,CONSISTENT QUALITY FILM AND INTERNAL KNITTED LINER. FULLY COATEDGAUNTLET. LENGTH 305MM. COLOUR BURGUNDY WITH BLACK HAND. 1 * 6 PAIR
Catalog # ANSE58-530/8
Supplier:  Ansell
GLOVES ALPHATEC. : LIQUID-PROOF NITRILE GLOVE WITH WET GRIP SURFACE,CONSISTENT QUALITY FILM AND INTERNAL KNITTED LINER. FULLY COATEDGAUNTLET. LENGTH 305MM. COLOUR BURGUNDY WITH BLACK HAND. 1 * 6 PAIR
Catalog # ANSE58-530/8
Supplier:  Ansell