Order Entry
China
Orders LinkContactUsLinkComponent
 

Drum funnels

260 mm, PP, 23 mm
Catalog # BURK9602-0250
Supplier:   BURKLE
Drum funnels
Catalog # BURK9602-0250
Supplier:  BURKLE
Drum funnels
Drum funnels

260 mm, PP, 23 mm

Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Made of PP, translucent.

  • Very good chemical resistance
  • Very good temperature resistance
  • Quick and easy to clean

For filling containers with large quantities of liquid or powdered substances quickly.

Specifications
  • Funnel O-Ø:
    260 mm
  • Material:
    PP
  • Seq:
    02
  • Stem O-Ø:
    23 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.