Drum funnels
Product Details & Documents
Made of PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
- Quick and easy to clean
For filling containers with large quantities of liquid or powdered substances quickly.
Documents
BURK9602-0250_TDS_EN
Datasheet
Specifications
- Funnel O-Ø:360 mm
- Material:PP
- Seq:03
- Stem O-Ø:31 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



