0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Inserts
Membrane inserts
Membrane inserts
Print
Membrane inserts
Membrane insert, red, 30 kD
Catalog # PALLOD030C65
Supplier:
CYTIVA PALL
Membrane inserts
Catalog # PALLOD030C65
Supplier:
CYTIVA PALL
Expand image
Return to Top
Membrane inserts
Membrane insert, red, 30 kD
Specifications
Add to cart
Jump to:
Specifications
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 12 件
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Product Literature
Pall Laboratory Centrifugal Devices Facilitate pure product with 90% recoveries in just minutes
Specifications
MWCO:
30 kD
Description:
Membrane insert, red
Seq:
03