Reversed phase HPLC columns, PRP-3
Semi-preparative, PRP-3, Stainless steel, 300 Å, 12 - 20 µm, 21,20 mm, 250 mm, PSDVB
目录编号 HAMI79469
供应商: HAMILTON BONADUZ
Jump to:
Product Details & Documents
PRP-3 polymeric reversed phase columns are ideal for gradient separations of peptides and proteins. The columns are packed with a poly(styrene-divinyl)benzene support.
- Better sample recovery than silica based columns
- Excellent durability
- Last longer than silica based columns
- Crosslinked to prevent shrinkage or swelling
Since the support is entirely polymeric and there are no C18 groups to hydrolyse, column lifetime is longer. Regardless of whether you use ionpair reagents or high percentage aqueous buffers (80 to 100%) for your separation, PRP-3 columns outlast silica based columns under common (pH 2,0) or uncommon (pH 12,7) conditions.
规格
- Column Format:Semi-preparative
- Ø int.:21,20 mm
- Particle Size:12 - 20 µm
- Pore size:300 Å
- Length:250 mm
- Seq:10
- Column hardware:Stainless steel
- Phase:PRP-3
- Packing material:PSDVB
- Cat. No.:HAMI79469
- Max. pressure:5000 psi
- USP code:L21
- Temperature limit:5 - 85 °C
- pH range:1 - 13



