Order Entry
China
ContactUsLinkComponent
Bottle, reagent, capacity 100ml, diameter 52mm, height 97mm, wide neck, with standard ground joint, NS 29/22, amber, soda lime glass 1 * 10 items
Catalog # SCOT231872406
Supplier:  DWK LIFE SCIENCES
undefined
Bottle, reagent, capacity 100ml, diameter 52mm, height 97mm, wide neck, with standard ground joint, NS 29/22, amber, soda lime glass 1 * 10 items
Catalog # SCOT231872406
Supplier:  DWK LIFE SCIENCES