Spatulas, micro
Straight spatula end, 150 mm, 4 mm
Catalog # RSGA531.004
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
- Micro spatulas with hexagon handle
- Heat and chemical resistant
- Choice of 2, 3 or 4 mm wide blade
Specifications
- Description:Straight spatula end
- Length:150 mm
- Blade width:4 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



